Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTCTCAGTTTGGTCATGTGGT[A/G]GACATTGTGGCTTTAAAGACCATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603520 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRPB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNRPB2 - small nuclear ribonucleoprotein polypeptide B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003092.4 | 250 | Silent Mutation | GTA,GTG | V,V 38 | NP_003083.1 | |
NM_198220.2 | 250 | Silent Mutation | GTA,GTG | V,V 38 | NP_937863.1 |