Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGACTCCACGCTGAACAGGTGA[A/G]TGTCCACACCGTGACGCGTGCCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608200 MIM: 601017 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK5RAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK5RAP1 - CDK5 regulatory subunit associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNTA1 - syntrophin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003098.2 | 1366 | Missense Mutation | ACT,ATT | T,I 378 | NP_003089.1 | |
XM_005260517.1 | 1366 | Missense Mutation | ACT,ATT | T,I 378 | XP_005260574.1 | |
XM_011529007.1 | 1366 | Missense Mutation | ACT,ATT | T,I 378 | XP_011527309.1 | |
XM_011529008.1 | 1366 | Missense Mutation | ACT,ATT | T,I 378 | XP_011527310.1 |