Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCAAGGCAGAAGCATCCGTTG[A/G]CAGCCAGATGCCCCGGTTTAAACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSM14B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSM14B - LSM family member 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144703.2 | 239 | Intron | NP_653304.2 | |||
XM_005260302.3 | 239 | Intron | XP_005260359.1 | |||
XM_011528605.2 | 239 | Intron | XP_011526907.1 | |||
XM_011528606.2 | 239 | Intron | XP_011526908.1 | |||
XM_011528607.2 | 239 | Intron | XP_011526909.1 | |||
XM_011528608.2 | 239 | Intron | XP_011526910.1 | |||
XM_011528609.2 | 239 | Intron | XP_011526911.1 | |||
XM_011528611.2 | 239 | Intron | XP_011526913.1 | |||
XM_011528612.2 | 239 | Intron | XP_011526914.1 | |||
XM_011528613.2 | 239 | Intron | XP_011526915.1 | |||
XM_011528614.2 | 239 | UTR 5 | XP_011526916.1 | |||
XM_011528615.2 | 239 | Intron | XP_011526917.1 | |||
XM_017027688.1 | 239 | Intron | XP_016883177.1 | |||
XM_017027689.1 | 239 | Intron | XP_016883178.1 | |||
XM_017027690.1 | 239 | Intron | XP_016883179.1 |