Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAACGGTTGTTTACAACCCTTA[C/T]GCTGCCCTTTCCATAGAGCAGCAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DHX35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX35 - DEAH-box helicase 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190809.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 42 | NP_001177738.1 | |
NM_021931.3 | 156 | Silent Mutation | TAC,TAT | Y,Y 42 | NP_068750.2 | |
XM_006723848.3 | 156 | Intron | XP_006723911.1 | |||
XM_011528972.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 42 | XP_011527274.1 | |
XM_011528974.1 | 156 | Silent Mutation | TAC,TAT | Y,Y 42 | XP_011527276.1 | |
XM_017028004.1 | 156 | Intron | XP_016883493.1 |