Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGCCTGCATAAATTTTCCACCC[A/G]GGATTATATCATGGAACCCTCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603169 MIM: 605297 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTSZ - cathepsin Z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFCD - negative elongation factor complex member C/D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198976.2 | 236 | Missense Mutation | CAG,CGG | Q,R 63 | NP_945327.2 |