Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTAGGTCAGCTGCTTGTGAGT[G/T]ATGGGGTATAGGGTAAGGCCAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCED1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCED1A - PC-esterase domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS16 - VPS16, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022575.3 | Intron | NP_072097.2 | ||||
NM_080413.2 | Intron | NP_536338.1 |