Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGGAGGGGAGCGGCCTCGGCTAC[A/C]TGGTGCAGGTGAAGCCCATGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGAP1 - ADP ribosylation factor GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COL20A1 - collagen type XX alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020882.2 | 286 | Missense Mutation | ATG,CTG | M,L 57 | NP_065933.2 | |
XM_011528937.1 | 286 | Missense Mutation | ATG,CTG | M,L 57 | XP_011527239.1 | |
XM_011528938.1 | 286 | Missense Mutation | ATG,CTG | M,L 57 | XP_011527240.1 | |
XM_011528939.1 | 286 | Missense Mutation | ATG,CTG | M,L 57 | XP_011527241.1 | |
XM_011528940.1 | 286 | Missense Mutation | ATG,CTG | M,L 57 | XP_011527242.1 |