Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATGACTTCCATGTCCGAGGACC[C/G]CCCCCAGTATATACCAACGGCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615717 MIM: 615718 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPIFB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BPIFB3 - BPI fold containing family B member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BPIFB4 - BPI fold containing family B member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182519.2 | 228 | Silent Mutation | CCC,CCG | P,P 71 | NP_872325.2 |