Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCCCAGCGGGAGGGTCTACGTG[C/T]TGAAGTTCAAGGCAGGGTCCAAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610650 MIM: 601033 MIM: 606731 | ||||||||||||||||||||
Literature Links: |
ADRM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADRM1 - adhesion regulating molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281437.1 | 1276 | Intron | NP_001268366.1 | |||
NM_001281438.1 | 1276 | Intron | NP_001268367.1 | |||
NM_007002.3 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | NP_008933.2 | |
NM_175573.2 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | NP_783163.1 | |
XM_005260257.1 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | XP_005260314.1 | |
XM_011528503.1 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | XP_011526805.1 | |
XM_017027602.1 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | XP_016883091.1 | |
XM_017027603.1 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | XP_016883092.1 | |
XM_017027604.1 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | XP_016883093.1 | |
XM_017027605.1 | 1276 | Silent Mutation | CTG,TTG | L,L 96 | XP_016883094.1 | |
XM_017027606.1 | 1276 | Intron | XP_016883095.1 | |||
XM_017027607.1 | 1276 | Intron | XP_016883096.1 | |||
XM_017027608.1 | 1276 | Intron | XP_016883097.1 | |||
XM_017027609.1 | 1276 | Intron | XP_016883098.1 | |||
XM_017027610.1 | 1276 | Intron | XP_016883099.1 |
LAMA5 - laminin subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSBPL2 - oxysterol binding protein like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |