Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTAGTGCTAGTGCAAATCAAAGA[C/G]AACTCTTATAAGCCAGAAATCGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609492 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASSF2 - Ras association domain family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014737.2 | 3152 | UTR 3 | NP_055552.1 | |||
NM_170774.1 | 3152 | UTR 3 | NP_739580.1 | |||
XM_005260895.3 | 3152 | UTR 3 | XP_005260952.1 | |||
XM_011529410.1 | 3152 | UTR 3 | XP_011527712.1 | |||
XM_011529411.1 | 3152 | UTR 3 | XP_011527713.1 | |||
XM_011529412.2 | 3152 | UTR 3 | XP_011527714.1 | |||
XM_017028149.1 | 3152 | UTR 3 | XP_016883638.1 | |||
XM_017028150.1 | 3152 | UTR 3 | XP_016883639.1 | |||
XM_017028151.1 | 3152 | UTR 3 | XP_016883640.1 | |||
XM_017028152.1 | 3152 | UTR 3 | XP_016883641.1 | |||
XM_017028153.1 | 3152 | UTR 3 | XP_016883642.1 |