Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCGTGTCTGTCCTCACCCTCCAG[A/C]CATGCTGAATCTATCCTCTTCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614108 MIM: 614110 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPIFB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BPIFB2 - BPI fold containing family B member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BPIFB6 - BPI fold containing family B member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174897.2 | Intron | NP_777557.1 | ||||
XM_017027663.1 | Intron | XP_016883152.1 |