Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCACCAGGACGCTGGCGCCCAGC[A/G]AGGTCAGCCGCGTCTCCTTCACGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147520 MIM: 610206 | ||||||||||||||||||||
Literature Links: |
ITPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITPA - inosine triphosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC4A11 - solute carrier family 4 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174089.1 | 2348 | Missense Mutation | TCG,TTG | S,L 742 | NP_001167560.1 | |
NM_001174090.1 | 2348 | Missense Mutation | TCG,TTG | S,L 785 | NP_001167561.1 | |
NM_032034.3 | 2348 | Missense Mutation | TCG,TTG | S,L 758 | NP_114423.1 | |
XM_005260856.4 | 2348 | Missense Mutation | TCG,TTG | S,L 865 | XP_005260913.1 | |
XM_005260857.1 | 2348 | Missense Mutation | TCG,TTG | S,L 723 | XP_005260914.1 | |
XM_011529383.2 | 2348 | Missense Mutation | TCG,TTG | S,L 731 | XP_011527685.1 | |
XM_011529384.1 | 2348 | Missense Mutation | TCG,TTG | S,L 723 | XP_011527686.1 | |
XM_011529385.1 | 2348 | Missense Mutation | TCG,TTG | S,L 723 | XP_011527687.1 | |
XM_017028093.1 | 2348 | Missense Mutation | TCG,TTG | S,L 863 | XP_016883582.1 | |
XM_017028094.1 | 2348 | Missense Mutation | TCG,TTG | S,L 723 | XP_016883583.1 | |
XM_017028095.1 | 2348 | Missense Mutation | TCG,TTG | S,L 704 | XP_016883584.1 | |
XM_017028096.1 | 2348 | Missense Mutation | TCG,TTG | S,L 723 | XP_016883585.1 | |
XM_017028097.1 | 2348 | Intron | XP_016883586.1 |