Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGGACCACGGGCCGGGTCAGG[C/T]CACCCAGCGAGGGGCTTGTATGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613979 | ||||||||||||||||||||
Literature Links: |
PRPF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRPF6 - pre-mRNA processing factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SAMD10 - sterile alpha motif domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080621.4 | 504 | Missense Mutation | GAC,GGC | D,G 110 | NP_542188.1 | |
XM_005260199.4 | 504 | Missense Mutation | GAC,GGC | D,G 149 | XP_005260256.1 | |
XM_006723705.3 | 504 | Missense Mutation | GAC,GGC | D,G 116 | XP_006723768.1 | |
XM_011528565.2 | 504 | Missense Mutation | GAC,GGC | D,G 314 | XP_011526867.1 | |
XM_017027671.1 | 504 | Missense Mutation | GAC,GGC | D,G 110 | XP_016883160.1 |
ZNF512B - zinc finger protein 512B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |