Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAGGCTGGGTTCTCTGAAGATG[A/G]TTACACGGCATTAATCTCCCAAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603006 | ||||||||||||||||||||
Literature Links: |
CDH4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH4 - cadherin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252338.2 | 204 | Intron | NP_001239267.1 | |||
NM_001252339.2 | 204 | Intron | NP_001239268.1 | |||
NM_001794.4 | 204 | Missense Mutation | GAT,GGT | D,G 39 | NP_001785.2 |