Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGACCATGGCAGTATTTCATGAC[A/G]AGGTGGAAATCGAGGACTTCCAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120270 MIM: 604745 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL9A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL9A3 - collagen type IX alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DPH3P1 - diphthamide biosynthesis 3 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080750.4 | Intron | NP_542788.1 |
TCFL5 - transcription factor like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301726.1 | Intron | NP_001288655.1 | ||||
NM_006602.3 | Intron | NP_006593.2 | ||||
XM_005260185.3 | Intron | XP_005260242.1 | ||||
XM_011528497.2 | Intron | XP_011526799.1 | ||||
XM_011528498.2 | Intron | XP_011526800.1 | ||||
XM_011528499.1 | Intron | XP_011526801.1 |