Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGTGCTGCTGGGAAATGCGGC[A/G]TGGTGAGCCTGGCTGTTGTTTAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 116957 | ||||||||||||||||||||
Literature Links: |
RBL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBL1 - RB transcriptional corepressor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323281.1 | 2927 | Missense Mutation | CGC,TGC | R,C 550 | NP_001310210.1 | |
NM_001323282.1 | 2927 | Missense Mutation | CGC,TGC | R,C 550 | NP_001310211.1 | |
NM_002895.4 | 2927 | Missense Mutation | CGC,TGC | R,C 977 | NP_002886.2 | |
NM_183404.3 | 2927 | Missense Mutation | CGC,TGC | R,C 977 | NP_899662.1 | |
XM_006723842.2 | 2927 | Missense Mutation | CGC,TGC | R,C 805 | XP_006723905.1 | |
XM_011528955.2 | 2927 | Intron | XP_011527257.1 | |||
XM_011528958.2 | 2927 | Intron | XP_011527260.1 | |||
XM_017027992.1 | 2927 | Missense Mutation | CGC,TGC | R,C 873 | XP_016883481.1 |