Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGCGTCTCCTTCACGTTCACAAT[C/G]CTGCGGTGGCCCGAGCCGCGAGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147520 MIM: 610206 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITPA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITPA - inosine triphosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC4A11 - solute carrier family 4 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174089.1 | 2316 | Silent Mutation | ACC,ACG | T,T 731 | NP_001167560.1 | |
NM_001174090.1 | 2316 | Silent Mutation | ACC,ACG | T,T 774 | NP_001167561.1 | |
NM_032034.3 | 2316 | Silent Mutation | ACC,ACG | T,T 747 | NP_114423.1 | |
XM_005260856.4 | 2316 | Silent Mutation | ACC,ACG | T,T 854 | XP_005260913.1 | |
XM_005260857.1 | 2316 | Silent Mutation | ACC,ACG | T,T 712 | XP_005260914.1 | |
XM_011529383.2 | 2316 | Silent Mutation | ACC,ACG | T,T 720 | XP_011527685.1 | |
XM_011529384.1 | 2316 | Silent Mutation | ACC,ACG | T,T 712 | XP_011527686.1 | |
XM_011529385.1 | 2316 | Silent Mutation | ACC,ACG | T,T 712 | XP_011527687.1 | |
XM_017028093.1 | 2316 | Silent Mutation | ACC,ACG | T,T 852 | XP_016883582.1 | |
XM_017028094.1 | 2316 | Silent Mutation | ACC,ACG | T,T 712 | XP_016883583.1 | |
XM_017028095.1 | 2316 | Silent Mutation | ACC,ACG | T,T 693 | XP_016883584.1 | |
XM_017028096.1 | 2316 | Silent Mutation | ACC,ACG | T,T 712 | XP_016883585.1 | |
XM_017028097.1 | 2316 | Intron | XP_016883586.1 |