Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAAGGTCATCAGGCACCACCAC[C/G]GGGTCTGGGGAGCCTGTGTGTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605020 | ||||||||||||||||||||
Literature Links: |
VSX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VSX1 - visual system homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256271.1 | 1161 | Missense Mutation | CGT,GGT | R,G 233 | NP_001243200.1 | |
NM_001256272.1 | 1161 | Missense Mutation | CCG,CGG | P,R 293 | NP_001243201.1 | |
NM_014588.5 | 1161 | Intron | NP_055403.2 | |||
NM_199425.2 | 1161 | Intron | NP_955457.1 | |||
XM_017027837.1 | 1161 | UTR 3 | XP_016883326.1 | |||
XM_017027838.1 | 1161 | UTR 3 | XP_016883327.1 |