Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGGGGCTCAACCCGGAGTGCAC[G/T]CCTTGCAACTCAAGCCCGTGTGCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607120 | ||||||||||||||||||||
Literature Links: |
PLCB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLCB1 - phospholipase C beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015192.3 | 415 | Missense Mutation | GCC,TCC | A,S 10 | NP_056007.1 | |
NM_182734.2 | 415 | Missense Mutation | GCC,TCC | A,S 10 | NP_877398.1 | |
XM_011529199.2 | 415 | Missense Mutation | GCC,TCC | A,S 10 | XP_011527501.1 | |
XM_011529200.2 | 415 | Intron | XP_011527502.1 | |||
XM_011529201.2 | 415 | Intron | XP_011527503.1 | |||
XM_011529203.2 | 415 | Intron | XP_011527505.1 | |||
XM_017027752.1 | 415 | Missense Mutation | GCC,TCC | A,S 10 | XP_016883241.1 | |
XM_017027753.1 | 415 | Intron | XP_016883242.1 |