Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAGGCTGCTCTCTGGGCGGGG[A/C]CACAGAGCCATGCACTCGCACGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613942 | ||||||||||||||||||||
Literature Links: |
SUN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SUN5 - Sad1 and UNC84 domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080675.3 | 1183 | Missense Mutation | NP_542406.2 | |||
XM_011528573.1 | 1183 | Missense Mutation | XP_011526875.1 | |||
XM_011528574.1 | 1183 | Missense Mutation | XP_011526876.1 | |||
XM_011528575.1 | 1183 | Missense Mutation | XP_011526877.1 | |||
XM_011528576.1 | 1183 | Intron | XP_011526878.1 |