Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTCTGCATCCTCATCTTGTCAC[A/G]GTATCCTCCCTGGGCCTGCAGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
WFDC10B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WFDC10B - WAP four-disulfide core domain 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172006.2 | 626 | Intron | NP_742003.1 | |||
NM_172131.2 | 626 | Intron | NP_742143.1 | |||
XM_017027824.1 | 626 | Missense Mutation | CGT,TGT | R,C 24 | XP_016883313.1 | |
XM_017027825.1 | 626 | Missense Mutation | CGT,TGT | R,C 24 | XP_016883314.1 |