Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAGAGCCTGGGCCTCCTCCCC[A/G]GGTTCTCCTGCACCTGAGATAAAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609314 | ||||||||||||||||||||
Literature Links: |
RSPH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSPH1 - radial spoke head 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286506.1 | 909 | Silent Mutation | CCC,CCT | P,P 209 | NP_001273435.1 | |
NM_080860.3 | 909 | Silent Mutation | CCC,CCT | P,P 247 | NP_543136.1 | |
XM_005261208.2 | 909 | Silent Mutation | CCC,CCT | P,P 178 | XP_005261265.1 | |
XM_011529786.1 | 909 | Silent Mutation | CCC,CCT | P,P 223 | XP_011528088.1 |