Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGAATGAGGTCCTCCACTAAAC[A/G]TTCTATAGCTTGCGTTATCTTTTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DNAJC28 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAJC28 - DnaJ heat shock protein family (Hsp40) member C28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040192.2 | 1053 | Missense Mutation | CGT,TGT | R,C 202 | NP_001035282.1 | |
NM_001320746.1 | 1053 | Missense Mutation | CGT,TGT | R,C 202 | NP_001307675.1 | |
NM_017833.4 | 1053 | Missense Mutation | CGT,TGT | R,C 202 | NP_060303.2 |
TMEM50B - transmembrane protein 50B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |