Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCTGTGGACCTGAGGTAGCAA[C/G]TAAACTCTTAGAGTTAATCAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603920 MIM: 611428 MIM: 182465 | ||||||||||||||||||||
Literature Links: |
CRYZL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRYZL1 - crystallin zeta like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DONSON - downstream neighbor of SON | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017613.3 | 1331 | Missense Mutation | CTT,GTT | L,V 422 | NP_060083.1 |
SON - SON DNA binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |