Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAGTGGAGAACTCGCGAGCACTC[A/G]GCCAGTGGTGCCTCCAATTTGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL18A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL18A1 - collagen type XVIII alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030582.3 | Intron | NP_085059.2 | ||||
NM_130444.2 | Intron | NP_569711.2 | ||||
NM_130445.3 | Intron | NP_569712.2 |
COL18A1-AS1 - COL18A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |