Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCACCAGGGTCCTTGGACAGGT[C/A]ATAGAGCAGCGGGGGCTCATGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607574 | ||||||||||||||||||||
Literature Links: |
ARSA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARSA - arylsulfatase A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000487.5 | 1563 | Missense Mutation | GAC,TAC | D,Y 432 | NP_000478.3 | |
NM_001085425.2 | 1563 | Missense Mutation | GAC,TAC | D,Y 432 | NP_001078894.2 | |
NM_001085426.2 | 1563 | Missense Mutation | GAC,TAC | D,Y 432 | NP_001078895.2 | |
NM_001085427.2 | 1563 | Missense Mutation | GAC,TAC | D,Y 432 | NP_001078896.2 | |
NM_001085428.2 | 1563 | Missense Mutation | GAC,TAC | D,Y 346 | NP_001078897.1 | |
XM_011530690.2 | 1563 | Missense Mutation | GAC,TAC | D,Y 346 | XP_011528992.1 | |
XM_011530691.2 | 1563 | UTR 3 | XP_011528993.1 | |||
XM_017028800.1 | 1563 | Missense Mutation | GAC,TAC | D,Y 470 | XP_016884289.1 |