Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCACAGTGACTTTGAACAGCTTCC[A/G]GATGATGTTGCCATCTCGGCCAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601488 | ||||||||||||||||||||
Literature Links: |
NCF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCF4 - neutrophil cytosolic factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000631.4 | 599 | Silent Mutation | CCA,CCG | P,P 17 | NP_000622.2 | |
NM_013416.3 | 599 | Silent Mutation | CCA,CCG | P,P 17 | NP_038202.2 | |
XM_017028808.1 | 599 | UTR 5 | XP_016884297.1 |