Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATTTGGAAGAATTCCCAGTAGA[C/T]TGAGGCTATCATTTTCCTCTACATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENTHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENTHD1 - ENTH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152512.3 | 1748 | Missense Mutation | AAT,AGT | N,S 489 | NP_689725.2 | |
XM_006724149.3 | 1748 | Missense Mutation | AAT,AGT | N,S 489 | XP_006724212.1 | |
XM_006724150.3 | 1748 | Missense Mutation | AAT,AGT | N,S 436 | XP_006724213.1 | |
XM_006724151.1 | 1748 | Missense Mutation | AAT,AGT | N,S 376 | XP_006724214.1 | |
XM_006724153.3 | 1748 | Intron | XP_006724216.1 | |||
XM_011529927.2 | 1748 | Missense Mutation | AAT,AGT | N,S 468 | XP_011528229.1 | |
XM_011529928.2 | 1748 | Missense Mutation | AAT,AGT | N,S 408 | XP_011528230.1 | |
XM_011529930.2 | 1748 | Missense Mutation | AAT,AGT | N,S 225 | XP_011528232.1 | |
XM_011529931.2 | 1748 | Intron | XP_011528233.1 |