Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGTTCCTCAGCAGGCCCTGCAT[C/G]CACCACAAGTCCGCCGGCACATCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPLA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNPLA5 - patatin like phospholipase domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177675.1 | 916 | Missense Mutation | TGC,TGG | C,W 261 | NP_001171146.1 | |
NM_138814.3 | 916 | Missense Mutation | TGC,TGG | C,W 375 | NP_620169.1 | |
XM_011529959.2 | 916 | Missense Mutation | TGC,TGG | C,W 235 | XP_011528261.1 |