Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGGGGGGCATCCAGAGAACAAC[C/T]GTAATCACTTCCTCCTTCACCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 170998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPARA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPARA - peroxisome proliferator activated receptor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001928.2 | Intron | NP_001001928.1 | ||||
NM_005036.4 | Intron | NP_005027.2 | ||||
XM_005261655.3 | Intron | XP_005261712.1 | ||||
XM_005261656.3 | Intron | XP_005261713.1 | ||||
XM_006724269.3 | Intron | XP_006724332.1 | ||||
XM_006724270.3 | Intron | XP_006724333.1 | ||||
XM_011530239.2 | Intron | XP_011528541.1 | ||||
XM_011530240.2 | Intron | XP_011528542.1 | ||||
XM_011530241.2 | Intron | XP_011528543.1 | ||||
XM_011530242.2 | Intron | XP_011528544.1 | ||||
XM_011530243.2 | Intron | XP_011528545.1 | ||||
XM_011530244.2 | Intron | XP_011528546.1 | ||||
XM_011530245.2 | Intron | XP_011528547.1 | ||||
XM_017028839.1 | Intron | XP_016884328.1 | ||||
XM_017028840.1 | Intron | XP_016884329.1 |