Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACCATTCTCAATTTAGAGTGTA[C/T]CTAAAATGTCATCTGTGCTGGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607588 MIM: 608082 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPIL2 - peptidylprolyl isomerase like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YPEL1 - yippee like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013313.4 | 1839 | UTR 3 | NP_037445.1 |