Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGGTGGTGTCCGCATCGTCGG[C/T]GGGAAGGCTGCACAGCATGGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102480 MIM: 606230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACR - acrosin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097.2 | 147 | Silent Mutation | GGC,GGT | G,G 45 | NP_001088.2 |
LOC105373100 - uncharacterized LOC105373100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHANK3 - SH3 and multiple ankyrin repeat domains 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |