Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCGGTGACTCCCTTGCGGCAC[A/G]TGGCAATGAGAGGCTGCGAGAGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602496 MIM: 180370 | ||||||||||||||||||||
Literature Links: |
MPST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPST - mercaptopyruvate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX33 - testis expressed 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TST - thiosulfate sulfurtransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270483.1 | 783 | Missense Mutation | NP_001257412.1 | |||
NM_003312.5 | 783 | Missense Mutation | NP_003303.2 |