Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCAGCACTCCAGGGCCAACGCC[A/G]AAACAAGAGGCACCCTCAGCCCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC116 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC116 - coiled-coil domain containing 116 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152612.2 | 497 | Missense Mutation | CAA,CGA | Q,R 62 | NP_689825.2 | |
XM_006724159.2 | 497 | Missense Mutation | CAA,CGA | Q,R 128 | XP_006724222.1 | |
XM_011529984.2 | 497 | Missense Mutation | CAA,CGA | Q,R 27 | XP_011528286.1 | |
XM_011529985.1 | 497 | Missense Mutation | CAA,CGA | Q,R 128 | XP_011528287.1 |
SDF2L1 - stromal cell derived factor 2 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YDJC - YdjC homolog (bacterial) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |