Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGGTCTACCTGTATGTCCTGGTC[A/G]ACTCAGGTAAGGGTCCTGCTTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UPK3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UPK3A - uroplakin 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167574.1 | 229 | Missense Mutation | AAC,GAC | N,D 68 | NP_001161046.1 | |
NM_006953.3 | 229 | Missense Mutation | AAC,GAC | N,D 68 | NP_008884.1 |