Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGAGTGCCTGGCAGGAGCCTGG[A/C]AGCACCACGGACCTGTTCATCCTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613383 MIM: 603692 MIM: 607422 | ||||||||||||||||||||
Literature Links: |
ANKRD54 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD54 - ankyrin repeat domain 54 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GALR3 - galanin receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003614.1 | 187 | Silent Mutation | GGA,GGC | G,G 54 | NP_003605.1 |
GCAT - glycine C-acetyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |