Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAAGAAGTAAAGCAGCTTCTGT[C/T]AATGTGACTTAAAACCATCTATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf42 - chromosome 22 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010859.1 | Intron | NP_001010859.1 | ||||
XM_011529922.2 | Intron | XP_011528224.1 | ||||
XM_017028629.1 | Intron | XP_016884118.1 | ||||
XM_017028630.1 | Intron | XP_016884119.1 | ||||
XM_017028631.1 | Intron | XP_016884120.1 | ||||
XM_017028632.1 | Intron | XP_016884121.1 |