Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGTTCTATTCTTTCTTTGATCT[C/T]TCGCTCTTCACGTTTTCGCTCATAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606796 | ||||||||||||||||||||
Literature Links: |
ST13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST13 - suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278589.1 | 1204 | Missense Mutation | AAG,GAG | K,E 236 | NP_001265518.1 | |
NM_003932.4 | 1204 | Missense Mutation | AAG,GAG | K,E 246 | NP_003923.2 |