Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAGGCAGCTCCGCCAGGCTACC[C/T]GGGGTGGCAAGGAGCCTCAGGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608926 MIM: 133450 | ||||||||||||||||||||
Literature Links: |
EMID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EMID1 - EMI domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EWSR1 - EWS RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHBDD3 - rhomboid domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006724224.3 | 953 | Silent Mutation | CCA,CCG | P,P 89 | XP_006724287.1 | |
XM_011530107.2 | 953 | Silent Mutation | CCA,CCG | P,P 217 | XP_011528409.1 | |
XM_017028749.1 | 953 | Silent Mutation | CCA,CCG | P,P 220 | XP_016884238.1 | |
XM_017028750.1 | 953 | Silent Mutation | CCA,CCG | P,P 84 | XP_016884239.1 |