Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCCGCAGCGTGGGCGCGGTGAG[A/G]ACGCGCAGCTCATGCAGCCGCTCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603721 | ||||||||||||||||||||
Literature Links: |
CCDC116 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC116 - coiled-coil domain containing 116 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2L3 - ubiquitin conjugating enzyme E2 L3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YDJC - YdjC homolog (bacterial) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017964.1 | 865 | Silent Mutation | GTC,GTT | V,V 276 | NP_001017964.1 | |
XM_011529906.2 | 865 | UTR 3 | XP_011528208.1 |