Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAACATCCAGCAGCACACACAGC[C/T]GCTGCAGCTGGTTGGTCAACAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612733 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THOC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THOC5 - THO complex 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002877.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 628 | NP_001002877.1 | |
NM_001002878.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 628 | NP_001002878.1 | |
NM_001002879.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 628 | NP_001002879.1 | |
NM_003678.4 | 2077 | Missense Mutation | CAG,CGG | Q,R 628 | NP_003669.4 | |
XM_005261797.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 639 | XP_005261854.1 | |
XM_005261798.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 639 | XP_005261855.1 | |
XM_005261799.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 639 | XP_005261856.1 | |
XM_005261801.3 | 2077 | Missense Mutation | CAG,CGG | Q,R 289 | XP_005261858.1 | |
XM_006724347.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 639 | XP_006724410.1 | |
XM_017029027.1 | 2077 | Missense Mutation | CAG,CGG | Q,R 639 | XP_016884516.1 |