Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGGACACCTTAGGCTTGGTGG[C/T]GGGGTCAGGTGGCGGCGGTTTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613619 | ||||||||||||||||||||
Literature Links: |
SCARF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCARF2 - scavenger receptor class F member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153334.6 | 2083 | Missense Mutation | NP_699165.3 | |||
NM_182895.4 | 2083 | Missense Mutation | NP_878315.2 | |||
XM_017029065.1 | 2083 | Intron | XP_016884554.1 |