Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTACACAATTTGACTTCTTCGC[G/T]TAGAGACTCCACACAGACAGCCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614785 MIM: 615404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFF - mitochondrial fission factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM4SF20 - transmembrane 4 L six family member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011511876.2 | 612 | Missense Mutation | AAG,ACG | K,T 156 | XP_011510178.1 |