Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACTGGCCAACATCAGGAGGGTT[C/T]CGGGAGGGTCTCAAGCCATACATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607334 | ||||||||||||||||||||
Literature Links: |
TRAK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRAK2 - trafficking kinesin protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015049.2 | 2816 | Silent Mutation | CGA,CGG | R,R 822 | NP_055864.2 | |
XM_011511690.1 | 2816 | Silent Mutation | CGA,CGG | R,R 822 | XP_011509992.1 | |
XM_017004772.1 | 2816 | Silent Mutation | CGA,CGG | R,R 611 | XP_016860261.1 |