Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCAGTGCGGGCAGGATGGCACG[C/T]TCCGCCACAGGTGGGCTCAGAAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC74B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC74B - coiled-coil domain containing 74B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258307.1 | Intron | NP_001245236.1 | ||||
NM_207310.2 | Intron | NP_997193.1 | ||||
XM_005263840.2 | Intron | XP_005263897.1 | ||||
XM_005263842.2 | Intron | XP_005263899.1 | ||||
XM_006712833.2 | Intron | XP_006712896.1 | ||||
XM_006712834.3 | Intron | XP_006712897.1 | ||||
XM_006712835.2 | Intron | XP_006712898.1 | ||||
XM_006712837.2 | Intron | XP_006712900.1 | ||||
XM_011512142.2 | Intron | XP_011510444.1 | ||||
XM_011512147.1 | Intron | XP_011510449.1 |
MED15P9 - mediator complex subunit 15 pseudogene 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |