Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGCCGCCTGGAAGGCTGACGGC[C/G]TGTGCTCCACCGTGGCCGGCAGTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606587 | ||||||||||||||||||||
Literature Links: |
PTPN18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTPN18 - protein tyrosine phosphatase, non-receptor type 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142370.1 | 185 | Intron | NP_001135842.1 | |||
NM_014369.3 | 185 | Missense Mutation | CTG,GTG | L,V 45 | NP_055184.2 | |
XM_006712416.3 | 185 | Intron | XP_006712479.1 | |||
XM_006712417.1 | 185 | Missense Mutation | CTG,GTG | L,V 45 | XP_006712480.1 | |
XM_017003798.1 | 185 | UTR 5 | XP_016859287.1 |