Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGATTCGTCCTTTATACATACC[A/G]TCAGAGCCCGTCTGTTCCTTTAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165640 | ||||||||||||||||||||
Literature Links: |
LOC101929715 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929715 - uncharacterized LOC101929715 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODC1 - ornithine decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287188.1 | 1473 | Silent Mutation | GAC,GAT | D,D 175 | NP_001274117.1 | |
NM_001287189.1 | 1473 | Silent Mutation | GAC,GAT | D,D 304 | NP_001274118.1 | |
NM_001287190.1 | 1473 | Silent Mutation | GAC,GAT | D,D 304 | NP_001274119.1 | |
NM_002539.2 | 1473 | Silent Mutation | GAC,GAT | D,D 304 | NP_002530.1 |
SNORA80B - small nucleolar RNA, H/ACA box 80B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |