Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTTGAGTTTCTTGAGATGTGTT[A/G]TCATTTTCACTTTGAAGTGAATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601912 | ||||||||||||||||||||
Literature Links: |
KIAA2012 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA2012 - KIAA2012 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUMO1 - small ubiquitin-like modifier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005781.1 | 195 | Missense Mutation | ACA,ATA | T,I 41 | NP_001005781.1 | |
NM_001005782.1 | 195 | Missense Mutation | ACA,ATA | T,I 16 | NP_001005782.1 | |
NM_003352.4 | 195 | Missense Mutation | ACA,ATA | T,I 41 | NP_003343.1 |