Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGATTCCAAAGCATGTACAGCCA[C/T]GGATGTTCTCCAGTCCTCTGCCGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603301 | ||||||||||||||||||||
Literature Links: |
CYS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYS1 - cystin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLF11 - Kruppel like factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177716.1 | 424 | Missense Mutation | ACG,ATG | T,M 119 | NP_001171187.1 | |
NM_001177718.1 | 424 | Missense Mutation | ACG,ATG | T,M 119 | NP_001171189.1 | |
NM_003597.4 | 424 | Missense Mutation | ACG,ATG | T,M 136 | NP_003588.1 |
LOC101929882 - uncharacterized LOC101929882 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |