Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTCCTCATTTCAGAAATTCC[A/G]GGTAGATATGCCTGGCTCAGGCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601575 | ||||||||||||||||||||
Literature Links: |
FLJ31356 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLJ31356 - uncharacterized protein FLJ31356 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FOSL2 - FOS like 2, AP-1 transcription factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005253.3 | 1072 | Missense Mutation | CAG,CGG | Q,R 37 | NP_005244.1 | |
XM_005264231.3 | 1072 | Missense Mutation | CAG,CGG | Q,R 37 | XP_005264288.1 | |
XM_006711976.2 | 1072 | Missense Mutation | CAG,CGG | Q,R 37 | XP_006712039.1 | |
XM_006711977.3 | 1072 | UTR 5 | XP_006712040.1 | |||
XM_017003737.1 | 1072 | UTR 5 | XP_016859226.1 |